Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mboat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083341.1 | NP_001076810.1 | 5 - 6 | 514 | 106 | |
NM_026037.3 | NP_080313.2 | 6 - 7 | 610 | 106 | ||
GenBank mRNA | AB297383.1 | - | 6 - 7 | 509 | 106 | |
AB305046.1 | - | 6 - 7 | 509 | 106 | ||
AK012931.1 | - | 5 - 6 | 498 | 106 | ||
AK031824.1 | - | 5 - 6 | 499 | 106 | ||
AK033106.1 | - | 3 - 4 | 274 | 106 | ||
AK034873.1 | - | 3 - 4 | 298 | 106 | ||
AK076045.1 | - | 5 - 6 | 504 | 106 | ||
AK215458.1 | - | 4 - 5 | 106 | |||
AU067514.1 | - | 6 - 7 | 610 | 106 | ||
BC025020.1 | - | 5 - 6 | 464 | 5364068 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |