Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290416.1 | NP_001277345.1 | 9 - 10 | 1065 | 75 | |
NM_026146.4 | NP_080422.1 | 9 - 10 | 971 | 75 | ||
GenBank mRNA | AK014556.1 | - | 9 - 10 | 1066 | 75 | |
AK019490.2 | - | 9 - 10 | 965 | 75 | ||
AY074931.1 | - | 8 - 9 | 908 | 75 | ||
BC027043.1 | - | 9 - 10 | 1255 | 75 | ||
BC100383.1 | - | 9 - 10 | 1153 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |