Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Eps8l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001290416.1 | NP_001277345.1 | 17 - 18 | 1913 | 72 | |
NM_026146.4 | NP_080422.1 | 17 - 18 | 1819 | 72 | ||
GenBank mRNA | AK019490.2 | - | 17 - 18 | 1813 | 72 | |
AY074931.1 | - | 16 - 17 | 1756 | 72 | ||
BC027043.1 | - | 17 - 18 | 2099 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |