Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rspry1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026274.4 | NP_080550.3 | 12 - 13 | 2019 | 67 | |
GenBank mRNA | AK013003.1 | - | 3 - 4 | 236 | 67 | |
AK031153.1 | - | 3 - 4 | 335 | 67 | ||
AK032416.1 | - | 12 - 13 | 1982 | 67 | ||
AK076799.1 | - | 12 - 13 | 1592 | 67 | ||
AK134544.1 | - | 9 - 10 | 886 | 67 | ||
AK141443.1 | - | 12 - 13 | 1569 | 67 | ||
AK150941.1 | - | 11 - 12 | 1494 | 67 | ||
AK151018.1 | - | 12 - 13 | 1989 | 67 | ||
AK152024.1 | - | 12 - 13 | 1989 | 67 | ||
AK152435.1 | - | 11 - 12 | 1494 | 67 | ||
AK172567.1 | - | 12 - 13 | 1985 | 67 | ||
AK173313.1 | - | 12 - 13 | 1638 | 67 | ||
BC010833.1 | - | 12 - 13 | 1634 | 4235165 | 67 | |
BC054121.1 | - | 12 - 13 | 2005 | 6393910 | 67 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |