Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc25a22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001177576.1 | NP_001171047.1 | 6 - 7 | 633 | 88 | |
NM_026646.3 | NP_080922.1 | 6 - 7 | 875 | 88 | ||
GenBank mRNA | AK010193.1 | - | 6 - 7 | 621 | 88 | |
AK018760.1 | - | 6 - 7 | 658 | 88 | ||
AK046874.1 | - | 6 - 7 | 620 | 88 | ||
AK048389.1 | - | 6 - 7 | 604 | 88 | ||
AK049313.1 | - | 6 - 7 | 604 | 88 | ||
AK155276.1 | - | 6 - 7 | 627 | 88 | ||
AK162400.1 | - | 6 - 7 | 586 | 88 | ||
BC037949.1 | - | 6 - 7 | 632 | 5367108 | 88 | |
BC050887.1 | - | 5 - 6 | 1351 | 88 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |