Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep57 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_026665.4 | NP_080941.3 | 6 - 7 | 907 | 104 | |
NR_037622.1 | 6 - 7 | 1167 | 104 | |||
GenBank mRNA | AK014862.1 | - | 3 - 4 | 540 | 104 | |
AK014873.1 | - | 6 - 7 | 854 | 104 | ||
AK014982.1 | - | 4 - 5 | 365 | 104 | ||
AK016484.1 | - | 6 - 7 | 691 | 104 | ||
AK019414.1 | - | 6 - 7 | 787 | 104 | ||
AK028458.1 | - | 6 - 7 | 735 | 104 | ||
AK129053.1 | - | 6 - 7 | 856 | 104 | ||
AK167523.1 | - | 6 - 7 | 792 | 104 | ||
AY225093.1 | - | 6 - 7 | 700 | 104 | ||
AY251192.1 | - | 6 - 7 | 852 | 104 | ||
BC025588.1 | - | 5 - 6 | 687 | 104 | ||
BC030353.1 | - | 6 - 7 | 1178 | 104 | ||
BC050785.1 | - | 4 - 5 | 418 | 6703205 | 104 | |
BC056968.1 | - | 6 - 7 | 1153 | 104 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |