Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Leng1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027203.3 | NP_081479.1 | 1 - 2 | 525 | 85 | |
GenBank mRNA | AK005095.1 | - | 1 - 2 | 156 | 85 | |
AK037634.1 | - | 1 - 2 | 172 | 85 | ||
BC023108.1 | - | 1 - 2 | 525 | 4021762 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |