Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zcchc8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027494.3 | NP_081770.3 | 8 - 9 | 862 | 92 | |
GenBank mRNA | AK017857.1 | - | 8 - 9 | 758 | 92 | |
AK032349.1 | - | 8 - 9 | 839 | 92 | ||
AK033469.1 | - | 8 - 9 | 812 | 92 | ||
AK156770.1 | - | 8 - 9 | 834 | 92 | ||
AK166990.1 | - | 8 - 9 | 769 | 92 | ||
AK167126.1 | - | 8 - 9 | 767 | 92 | ||
AK167736.1 | - | 8 - 9 | 769 | 92 | ||
BC013555.1 | - | 7 - 8 | 514 | 4238634 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |