Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0900003860.mm.1 |
NM_027498.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003860.mm.1 |
NM_027498.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003860.mm.1 |
TC0900000550.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003867.mm.1 |
NM_027498.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003867.mm.1 |
NM_027498.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003867.mm.1 |
TC0900000550.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003864.mm.1 |
NM_027498.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003864.mm.1 |
NM_027498.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003864.mm.1 |
TC0900000550.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003869.mm.1 |
NM_027498.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003869.mm.1 |
NM_027498.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003869.mm.1 |
TC0900000550.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0900003862.mm.1 |
NM_027498.3.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003862.mm.1 |
NM_027498.3 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0900003862.mm.1 |
TC0900000550.mm.1 |
MTA TC |