Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rufy3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001289774.1 | NP_001276703.1 | 8 - 9 | 1153 | 138 | |
NM_001289775.1 | NP_001276704.1 | 8 - 9 | 1153 | 138 | ||
NM_001289776.1 | NP_001276705.1 | 9 - 10 | 1575 | 138 | ||
NM_001289777.1 | NP_001276706.1 | 10 - 11 | 1400 | 138 | ||
NM_027530.3 | NP_081806.1 | 8 - 9 | 1521 | 138 | ||
GenBank mRNA | AK013185.1 | - | 10 - 11 | 1401 | 138 | |
AK018184.1 | - | 8 - 9 | 1499 | 138 | ||
AK083242.1 | - | 8 - 9 | 1456 | 138 | ||
AK148795.1 | - | 8 - 9 | 817 | 138 | ||
AK154941.1 | - | 8 - 9 | 1154 | 138 | ||
BC017648.1 | - | 8 - 9 | 910 | 4527858 | 138 | |
BC049127.1 | - | 8 - 9 | 1460 | 6821567 | 138 | |
BC058259.1 | - | 9 - 10 | 1294 | 6469137 | 138 | |
CT010404.1 | - | 8 - 9 | 894 | 138 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |