Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmrtc1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027591.3 | NP_081867.1 | 1 - 2 | 74 | 68 | |
GenBank mRNA | AK014934.1 | - | 1 - 2 | 75 | 68 | |
BC147175.1 | - | 1 - 2 | 46 | 9056223 | 68 | |
BC147176.1 | - | 1 - 2 | 46 | 9056216 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |