Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tchhl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_027762.3 | NP_082038.2 | 1 - 2 | 59 | 61 | |
GenBank mRNA | AK017244.1 | - | 1 - 2 | 60 | 61 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mus musculus Probe spans exons |