Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dhx34 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001285931.1 | NP_001272860.1 | 3 - 4 | 1822 | 110 | |
NM_001285932.1 | NP_001272861.1 | 3 - 4 | 1588 | 110 | ||
NM_027883.3 | NP_082159.3 | 3 - 4 | 1313 | 110 | ||
GenBank mRNA | AK004733.1 | - | 3 - 4 | 1314 | 110 | |
AK076217.1 | - | 2 - 3 | 1288 | 110 | ||
AK129062.1 | - | 3 - 4 | 110 | |||
BC058370.1 | - | 3 - 4 | 1650 | 110 | ||
BC080856.1 | - | 3 - 4 | 1588 | 6854931 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |