Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmco6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028036.3 | NP_082312.3 | 5 - 6 | 703 | 57 | |
GenBank mRNA | AK010487.1 | - | 5 - 6 | 628 | 57 | |
AK046207.1 | - | 5 - 6 | 741 | 57 | ||
BC005637.1 | - | 5 - 6 | 705 | 3156734 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |