Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fundc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028058.3 | NP_082334.1 | 1 - 2 | 145 | 109 | |
GenBank mRNA | AK005163.1 | - | 1 - 2 | 146 | 109 | |
AK168868.1 | - | 1 - 2 | 132 | 109 | ||
BC012515.1 | - | 1 - 2 | 87 | 2651125 | 109 | |
BC030487.1 | - | 1 - 2 | 151 | 5374103 | 109 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |