Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hspa12b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028306.3 | NP_082582.1 | 8 - 9 | 975 | 60 | |
GenBank mRNA | AK012548.1 | - | 8 - 9 | 948 | 60 | |
AY196790.1 | - | 7 - 8 | 855 | 60 | ||
BC011103.1 | - | 8 - 9 | 975 | 3990088 | 60 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |