Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mon1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028369.3 | NP_082645.1 | 4 - 5 | 1670 | 92 | |
GenBank mRNA | AK013387.1 | - | 4 - 5 | 1617 | 92 | |
AK172624.1 | - | 4 - 5 | 1577 | 92 | ||
AK211446.1 | - | 1 - 2 | 102 | 92 | ||
BC006070.1 | - | 1 - 2 | 477 | 92 | ||
BC048852.1 | - | 4 - 5 | 1596 | 5148425 | 92 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |