Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028716.4 | NP_082992.1 | 8 - 9 | 816 | 75 | |
GenBank mRNA | AK014030.1 | - | 7 - 8 | 1236 | 75 | |
AK014380.1 | - | 8 - 9 | 817 | 75 | ||
BC027776.1 | - | 8 - 9 | 811 | 5352251 | 75 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |