Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028766.2 | NP_083042.1 | 3 - 4 | 434 | 99 | |
GenBank mRNA | AK004778.1 | - | 3 - 4 | 435 | 99 | |
AK037165.1 | - | 3 - 4 | 406 | 99 | ||
AK050173.1 | - | 3 - 4 | 473 | 99 | ||
AK151693.1 | - | 3 - 4 | 427 | 99 | ||
AK151766.1 | - | 3 - 4 | 452 | 99 | ||
AK152078.1 | - | 3 - 4 | 427 | 99 | ||
AK168559.1 | - | 3 - 4 | 442 | 99 | ||
BC005521.1 | - | 3 - 4 | 399 | 99 | ||
BC024933.1 | - | 3 - 4 | 421 | 3990353 | 99 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus 1700 Probe spans exons |