Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sash3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028773.3 | NP_083049.1 | 1 - 2 | 193 | 82 | |
GenBank mRNA | AJ306422.1 | - | 1 - 2 | 59 | 82 | |
AK004734.2 | - | 1 - 2 | 194 | 82 | ||
AK156202.1 | - | 1 - 2 | 258 | 82 | ||
AK171017.1 | - | 1 - 2 | 191 | 82 | ||
BC064734.1 | - | 1 - 2 | 178 | 82 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus 1700 Probe spans exons |