Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cep97 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_028815.4 | NP_083091.1 | 7 - 8 | 948 | 68 | |
GenBank mRNA | AK012971.2 | - | 7 - 8 | 933 | 68 | |
AK016556.1 | - | 7 - 8 | 1023 | 68 | ||
AK046486.1 | - | 7 - 8 | 1497 | 68 | ||
AK154885.1 | - | 7 - 8 | 927 | 68 | ||
BC067049.1 | - | 7 - 8 | 948 | 30363673 | 68 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |