Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tc2n | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082976.2 | NP_001076445.1 | 3 - 4 | 464 | 144 | |
NM_001286364.1 | NP_001273293.1 | 3 - 4 | 1127 | 144 | ||
NM_028924.3 | NP_083200.1 | 3 - 4 | 816 | 144 | ||
GenBank mRNA | AB062282.1 | - | 2 - 3 | 296 | 144 | |
AK016683.1 | - | 3 - 4 | 1148 | 144 | ||
AK136558.1 | - | 3 - 4 | 465 | 144 | ||
BC120793.1 | - | 3 - 4 | 816 | 40129716 | 144 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |