Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spag16 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271533.1 | NP_001258462.1 | 7 - 8 | 776 | 89 | |
NM_029160.3 | NP_083436.2 | 7 - 8 | 776 | 89 | ||
GenBank mRNA | AF490391.1 | - | 7 - 8 | 758 | 89 | |
AK015884.1 | - | 7 - 8 | 777 | 89 | ||
AK083083.1 | - | 7 - 8 | 755 | 89 | ||
BC120668.1 | - | 7 - 8 | 775 | 40129591 | 89 | |
BC145006.1 | - | 7 - 8 | 775 | 9053554 | 89 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Mus musculus Probe spans exons |