Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Usp50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029163.3 | NP_083439.2 | 4 - 5 | 832 | 72 | |
GenBank mRNA | AK015766.1 | - | 4 - 5 | 817 | 72 | |
BC061020.1 | - | 4 - 5 | 809 | 6772471 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Protease Mammalian Gene Collection Mus musculus Probe spans exons |