Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Phf14 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001168382.1 | NP_001161854.1 | 10 - 11 | 2377 | 86 | |
NM_029404.3 | NP_083680.2 | 10 - 11 | 2374 | 86 | ||
GenBank mRNA | AK003209.1 | - | 3 - 4 | 463 | 86 | |
AK016517.1 | - | 10 - 11 | 2377 | 86 | ||
AK173024.1 | - | 2 - 3 | 216 | 86 | ||
BC040236.1 | - | 10 - 11 | 2340 | 3585881 | 86 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |