Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Klhl24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_029436.3 | NP_083712.4 | 5 - 6 | 1466 | 66 | |
GenBank mRNA | AK015239.1 | - | 5 - 6 | 1452 | 66 | |
AK044892.1 | - | 5 - 6 | 1438 | 66 | ||
AK046446.1 | - | 3 - 4 | 544 | 66 | ||
AK163616.1 | - | 3 - 4 | 604 | 66 | ||
BC021407.1 | - | 3 - 4 | 1020 | 66 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |