Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Glt8d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001165930.1 | NP_001159402.1 | 7 - 8 | 720 | 113 | |
NM_029626.2 | NP_083902.2 | 7 - 8 | 1249 | 113 | ||
GenBank mRNA | AK010388.1 | - | 7 - 8 | 978 | 113 | |
AK077388.1 | - | 7 - 8 | 713 | 113 | ||
BC029215.1 | - | 5 - 6 | 923 | 113 | ||
BC055709.1 | - | 6 - 7 | 1136 | 113 | ||
BC058731.1 | - | 6 - 7 | 1136 | 113 | ||
BC069873.1 | - | 7 - 8 | 799 | 6314675 | 113 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |