Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aacs | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030210.1 | NP_084486.1 | 10 - 11 | 1233 | 74 | |
GenBank mRNA | AB054122.1 | - | 10 - 11 | 1191 | 74 | |
AB056114.1 | - | 10 - 11 | 1128 | 74 | ||
AK019063.1 | - | 10 - 11 | 1233 | 74 | ||
AK034184.1 | - | 10 - 11 | 1166 | 74 | ||
AK043848.1 | - | 10 - 11 | 1256 | 74 | ||
AK044017.1 | - | 10 - 11 | 1233 | 74 | ||
AK147189.1 | - | 10 - 11 | 1233 | 74 | ||
AK150266.1 | - | 10 - 11 | 1233 | 74 | ||
AK165418.1 | - | 10 - 11 | 1233 | 74 | ||
AK165457.1 | - | 10 - 11 | 1233 | 74 | ||
AK170120.1 | - | 10 - 11 | 1233 | 74 | ||
AK170653.1 | - | 10 - 11 | 1233 | 74 | ||
AK170856.1 | - | 10 - 11 | 1233 | 74 | ||
AK218938.1 | - | 2 - 3 | 154 | 74 | ||
BC026817.1 | - | 10 - 11 | 1224 | 4235112 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |