Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Amhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144547.2 | NP_653130.2 | 6 - 7 | 945 | 69 | |
GenBank mRNA | AF503863.1 | - | 6 - 7 | 892 | 69 | |
BC119571.1 | - | 6 - 7 | 876 | 40057985 | 69 | |
BC119572.2 | - | 5 - 6 | 69 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Protein Kinase Mammalian Gene Collection Mus musculus Probe spans exons |