Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Spats2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_144882.4 | NP_659131.2 | 3 - 4 | 301 | 74 | |
GenBank mRNA | AK012791.1 | - | 3 - 4 | 275 | 74 | |
AK019292.1 | - | 3 - 4 | 275 | 74 | ||
AK031266.1 | - | 3 - 4 | 256 | 74 | ||
AK032169.1 | - | 3 - 4 | 236 | 74 | ||
AK161988.1 | - | 3 - 4 | 274 | 74 | ||
BC021879.1 | - | 3 - 4 | 237 | 4216816 | 74 | |
EF410080.1 | - | 341 | 74 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |