Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Angptl6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145154.2 | NP_660136.1 | 3 - 4 | 844 | 58 | |
GenBank mRNA | AB054065.1 | - | 3 - 4 | 844 | 58 | |
AK018113.1 | - | 2 - 3 | 213 | 58 | ||
BC025904.1 | - | 3 - 4 | 822 | 5049765 | 58 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |