Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cald1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145575.3 | NP_663550.1 | 4 - 5 | 945 | 85 | |
GenBank mRNA | AF439859.1 | - | 3 - 4 | 890 | 85 | |
AK148009.1 | - | 2 - 3 | 317 | 85 | ||
BC019435.1 | - | 4 - 5 | 904 | 5148205 | 85 | |
BQ769983.1 | - | 1 - 2 | 129 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |