Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ranbp10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145824.4 | NP_665823.2 | 3 - 4 | 562 | 85 | |
GenBank mRNA | AK173177.1 | - | 3 - 4 | 375 | 85 | |
AY337314.1 | - | 3 - 4 | 563 | 85 | ||
BC024698.1 | - | 3 - 4 | 407 | 3667000 | 85 | |
EU281316.1 | - | 3 - 4 | 423 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Mus musculus Probe spans exons |