Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ccdc25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145944.4 | NP_666056.1 | 5 - 6 | 400 | 72 | |
GenBank mRNA | AK012172.1 | - | 5 - 6 | 348 | 72 | |
AK016286.1 | - | 5 - 6 | 312 | 72 | ||
AK078345.1 | - | 5 - 6 | 401 | 72 | ||
AK089982.1 | - | 5 - 6 | 325 | 72 | ||
AK139180.1 | - | 4 - 5 | 2776 | 72 | ||
BC025545.1 | - | 348 | 72 | |||
BC128015.1 | - | 5 - 6 | 292 | 40136066 | 72 | |
BC148235.1 | - | 5 - 6 | 299 | 40136084 | 72 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Mus musculus Probe spans exons |