Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csnk1g3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_152809.2 | NP_690022.2 | 7 - 8 | 1490 | 85 | |
GenBank mRNA | AK032675.1 | - | 1 - 2 | 45 | 85 | |
BC033601.1 | - | 4 - 5 | 505 | 85 | ||
BQ443928.1 | - | 4 - 5 | 524 | 85 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |