Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dtnb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001162465.1 | NP_001155937.1 | 11 - 12 | 1427 | 57 | |
NM_007886.2 | NP_031912.2 | 11 - 12 | 1362 | 57 | ||
NR_027869.1 | 11 - 12 | 1427 | 57 | |||
NR_027870.1 | 11 - 12 | 1427 | 57 | |||
GenBank mRNA | AJ003007.1 | - | 11 - 12 | 1295 | 57 | |
AK043436.1 | - | 11 - 12 | 1385 | 57 | ||
BC030924.1 | - | 11 - 12 | 1362 | 4910166 | 57 | |
Y15742.1 | - | 11 - 12 | 1259 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Mus musculus Probe spans exons |