Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Mecom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_007963.2 | NP_031989.2 | 5 - 6 | 1268 | 93 | |
GenBank mRNA | AJ001482.1 | - | 3 - 4 | 424 | 93 | |
AK033517.1 | - | 5 - 6 | 702 | 93 | ||
BC076620.1 | - | 5 - 6 | 1220 | 93 | ||
BC139762.1 | - | 3 - 4 | 386 | 40109903 | 93 | |
JQ665270.1 | - | 3 - 4 | 416 | 93 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |