Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Flot2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040403.1 | NP_001035493.1 | 7 - 8 | 824 | 63 | |
NM_001284227.1 | NP_001271156.1 | 8 - 9 | 908 | 63 | ||
NM_001284228.1 | NP_001271157.1 | 8 - 9 | 771 | 63 | ||
NM_008028.2 | NP_032054.1 | 8 - 9 | 915 | 63 | ||
GenBank mRNA | AK004589.1 | - | 7 - 8 | 762 | 63 | |
AK030199.1 | - | 7 - 8 | 787 | 63 | ||
AK149530.1 | - | 8 - 9 | 846 | 63 | ||
AK170557.1 | - | 7 - 8 | 825 | 63 | ||
AK208907.1 | - | 1 - 2 | 123 | 63 | ||
BC070423.1 | - | 8 - 9 | 873 | 6403816 | 63 | |
U07890.1 | - | 8 - 9 | 785 | 63 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |