Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dnajc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_008929.3 | NP_032955.2 | 6 - 7 | 888 | 70 | |
GenBank mRNA | AK004033.1 | - | 1 - 2 | 189 | 70 | |
AK146097.1 | - | 6 - 7 | 852 | 70 | ||
AK148270.1 | - | 6 - 7 | 845 | 70 | ||
BC013766.1 | - | 6 - 7 | 833 | 2646147 | 70 | |
U28423.1 | - | 6 - 7 | 831 | 70 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |