Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brca1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009764.3 | NP_033894.3 | 4 - 5 | 422 | 101 | |
GenBank mRNA | AB221611.1 | - | 3 - 4 | 216 | 101 | |
AK144702.1 | - | 4 - 5 | 341 | 101 | ||
BC068303.1 | - | 4 - 5 | 258 | 30431022 | 101 | |
U31625.1 | - | 4 - 5 | 279 | 101 | ||
U32446.1 | - | 4 - 5 | 332 | 101 | ||
U35641.1 | - | 4 - 5 | 315 | 101 | ||
U36475.1 | - | 3 - 4 | 242 | 101 | ||
U68174.1 | - | 4 - 5 | 326 | 101 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |