Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clgn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_009904.2 | NP_034034.2 | 12 - 13 | 1707 | 102 | |
GenBank mRNA | AK019534.1 | - | 12 - 13 | 1621 | 102 | |
AK034525.1 | - | 12 - 13 | 1608 | 102 | ||
BC050767.1 | - | 12 - 13 | 1707 | 6771060 | 102 | |
D14117.1 | - | 12 - 13 | 1720 | 102 | ||
D86323.1 | - | 12 - 13 | 1720 | 102 | ||
U08373.1 | - | 12 - 13 | 1720 | 102 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |