Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbln1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010180.2 | NP_034310.2 | 6 - 7 | 773 | 57 | |
GenBank mRNA | AK035388.1 | - | 6 - 7 | 726 | 57 | |
AK083573.1 | - | 6 - 7 | 776 | 57 | ||
AK085989.1 | - | 6 - 7 | 715 | 57 | ||
AK086451.1 | - | 6 - 7 | 774 | 57 | ||
AK137511.1 | - | 6 - 7 | 716 | 57 | ||
AK159646.1 | - | 6 - 7 | 728 | 57 | ||
EU543224.1 | - | 6 - 7 | 711 | 57 | ||
X70854.1 | - | 6 - 7 | 770 | 57 |
Set Membership: |
Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Mus musculus Probe spans exons |