Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Il18rap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_010553.4 | NP_034683.1 | 2 - 3 | 189 | 106 | |
GenBank mRNA | AF077347.1 | - | 2 - 3 | 198 | 106 | |
AK053176.1 | - | 2 - 3 | 104 | 106 | ||
AK137541.1 | - | 2 - 3 | 150 | 106 | ||
BC120598.1 | - | 2 - 3 | 196 | 40129521 | 106 | |
BC120600.1 | - | 2 - 3 | 196 | 40129523 | 106 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |