Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kifc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001145831.1 | NP_001139303.1 | 19 - 20 | 2656 | 71 | |
NM_001145832.1 | NP_001139304.1 | 15 - 16 | 2170 | 71 | ||
NM_010631.2 | NP_034761.3 | 17 - 18 | 2513 | 71 | ||
GenBank mRNA | AF013118.2 | - | 13 - 14 | 2149 | 71 | |
AK132895.1 | - | 15 - 16 | 2170 | 71 | ||
AK135926.1 | - | 12 - 13 | 1712 | 71 | ||
AK166482.1 | - | 19 - 20 | 2637 | 71 | ||
BC004069.1 | - | 9 - 10 | 1261 | 71 | ||
BC016118.1 | - | 12 - 13 | 1717 | 71 | ||
BC023374.1 | - | 10 - 11 | 1305 | 71 | ||
BC070429.1 | - | 17 - 18 | 2513 | 6810893 | 71 | |
D49546.1 | - | 17 - 18 | 2528 | 71 |
Set Membership: |
Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |