Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Mouse Clariom S Array |
PSR0100021410.mm.1 |
NM_001278075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021410.mm.1 |
TC0100002631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021415.mm.1 |
NM_001278075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021415.mm.1 |
NM_011811.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021415.mm.1 |
NR_102757.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021415.mm.1 |
NR_102758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021415.mm.1 |
TC0100002631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021412.mm.1 |
NM_011811.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021412.mm.1 |
NR_102757.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021412.mm.1 |
NR_102758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021412.mm.1 |
TC0100002631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021413.mm.1 |
NM_011811.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021413.mm.1 |
NR_102757.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021413.mm.1 |
NR_102758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021413.mm.1 |
TC0100002631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021414.mm.1 |
NM_001278075.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021414.mm.1 |
NM_011811.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021414.mm.1 |
NR_102757.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021414.mm.1 |
NR_102758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021414.mm.1 |
TC0100002631.mm.1 |
MTA TC |
Affymetrix Mouse Clariom S Array |
PSR0100021411.mm.1 |
NM_011811.4 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021411.mm.1 |
NR_102757.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021411.mm.1 |
NR_102758.1 |
Refseq |
Affymetrix Mouse Clariom S Array |
PSR0100021411.mm.1 |
TC0100002631.mm.1 |
MTA TC |