Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Casp8ap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001122978.1 | NP_001116450.1 | 7 - 8 | 2655 | 90 | |
NM_011997.2 | NP_036127.2 | 7 - 8 | 2902 | 90 | ||
GenBank mRNA | AF132726.1 | - | 7 - 8 | 2862 | 90 | |
BC138022.1 | - | 6 - 7 | 2645 | 8861041 | 90 | |
BC138041.1 | - | 6 - 7 | 2645 | 8861060 | 90 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Apoptosis Mammalian Gene Collection Mus musculus Probe spans exons |