Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ctbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001198861.1 | NP_001185790.1 | 9 - 10 | 1521 | 111 | |
NM_013502.3 | NP_038530.1 | 8 - 9 | 1328 | 111 | ||
GenBank mRNA | AB033122.1 | - | 8 - 9 | 1293 | 111 | |
AK133816.1 | - | 9 - 10 | 1492 | 111 | ||
AK165276.1 | - | 9 - 10 | 1476 | 111 | ||
AK170133.1 | - | 7 - 8 | 1035 | 111 | ||
AK171650.1 | - | 7 - 8 | 1161 | 111 | ||
AK210217.1 | - | 1 - 2 | 105 | 111 | ||
AK213613.1 | - | 1 - 2 | 69 | 111 | ||
BC013702.1 | - | 8 - 9 | 1241 | 3155102 | 111 | |
BC015071.1 | - | 8 - 9 | 1297 | 4192052 | 111 | |
BC042425.1 | - | 9 - 10 | 1502 | 5005681 | 111 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Mus musculus Probe spans exons |