Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ksr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_013571.2 | NP_038599.1 | 17 - 18 | 2481 | 110 | |
GenBank mRNA | AK137532.1 | - | 16 - 17 | 2218 | 110 | |
U43585.1 | - | 17 - 18 | 2488 | 110 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Protein Kinase Mus musculus Probe spans exons |