Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ercc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015769.2 | NP_056584.2 | 4 - 5 | 991 | 95 | |
GenBank mRNA | AF189285.1 | - | 4 - 5 | 991 | 95 | |
AK043366.1 | - | 4 - 5 | 766 | 95 | ||
AK138588.1 | - | 4 - 5 | 785 | 95 | ||
AK149326.1 | - | 4 - 5 | 824 | 95 | ||
BC026792.1 | - | 4 - 5 | 782 | 4911382 | 95 | |
BC051036.1 | - | 3 - 4 | 522 | 95 |
Set Membership: |
Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Mus musculus Probe spans exons |