Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxo18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_015792.1 | NP_056607.1 | 9 - 10 | 1659 | 107 | |
GenBank mRNA | AF184275.1 | - | 7 - 8 | 1153 | 107 | |
AK083118.1 | - | 9 - 10 | 2004 | 107 | ||
AK084645.1 | - | 9 - 10 | 1595 | 107 | ||
AK147829.1 | - | 9 - 10 | 1643 | 107 | ||
AK154357.1 | - | 9 - 10 | 1682 | 107 | ||
AK158937.1 | - | 9 - 10 | 1710 | 107 | ||
BC004036.1 | - | 6 - 7 | 745 | 107 | ||
BC031393.1 | - | 9 - 10 | 1659 | 4948804 | 107 |
Set Membership: |
Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Mus musculus Probe spans exons |